PRI424060702 Princeton BioMolecular Research, Inc.

1-Methyl-5-nitro-3-trifluoromethyl-1H-pyrazole

  • MDL number MFCD09701889
  • Cas Number : 1170473-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart