PRI424060921 Princeton BioMolecular Research, Inc.

(4-Iodo-1-methyl-1H-pyrazol-3-yl)-acetic acid

  • MDL number MFCD24539553
  • Cas Number : 1354705-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart