PRI424060944 Princeton BioMolecular Research, Inc.

4-Iodo-2-methyl-5-nitro-2H-pyrazole-3-carbonitrile

  • MDL number MFCD24539568
  • Cas Number : 1354703-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart