PRI424061001 Princeton BioMolecular Research, Inc.

1-(2,2-Difluoro-ethyl)-4-iodo-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD24539616
  • Cas Number : 1354706-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart