PRI424061033 Princeton BioMolecular Research, Inc.

1-Ethyl-4-iodo-5-nitro-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD24539644
  • Cas Number : 1354706-75-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart