PRI424061071 Princeton BioMolecular Research, Inc.

1-Methyl-4-(1-methyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid

  • MDL number MFCD24540299
  • Cas Number : 1354704-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart