PRI424061117 Princeton BioMolecular Research, Inc.

3-Bromo-1-(2,2,2-trifluoro-ethyl)-1H-pyrazole

  • MDL number MFCD24540334
  • Cas Number : 1354706-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart