PRI424061136 Princeton BioMolecular Research, Inc.

1-Isopropyl-5-methyl-1H-pyrazole-3-sulfonyl chloride

  • MDL number MFCD24540343
  • Cas Number : 1354704-57-6
  • Molecular Weight : 222.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart