PRI424061156 Princeton BioMolecular Research, Inc.

4-(1,3-Dimethyl-1H-pyrazol-4-yl)-1-methyl-1H-pyrazolo[3,4-b]pyridine-3-carboxylic acid

  • MDL number MFCD24540363
  • Cas Number : 1354706-19-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart