PRI424061179 Princeton BioMolecular Research, Inc.

3-(4-Ethynyl-pyrazol-1-yl)-propionic acid

  • MDL number MFCD24540381
  • Cas Number : 1354704-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart