PRI424061226 Princeton BioMolecular Research, Inc.

5-Cyclopropyl-4-ethynyl-1-methyl-1H-pyrazole

  • MDL number MFCD24540415
  • Cas Number : 1354706-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart