PRI424061326 Princeton BioMolecular Research, Inc.

1-Methyl-5-nitro-1H-pyrazole-3-carbonyl chloride

  • MDL number MFCD24540512
  • Cas Number : 1429418-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart