PRI424061347 Princeton BioMolecular Research, Inc.

3-(2,2-Difluoro-ethoxy)-1-isopropyl-4-nitro-1H-pyrazole

  • MDL number MFCD24540538
  • Cas Number : 1429418-58-5
  • Molecular Weight : 235.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart