PRI424061354 Princeton BioMolecular Research, Inc.

5-Fluoro-thiophene-2-carbonyl chloride

  • MDL number MFCD13173673
  • Cas Number : 207849-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart