PRI424061467 Princeton BioMolecular Research, Inc.

[1-(2-Fluoro-ethyl)-1H-pyrazol-3-yl]-acetonitrile

  • MDL number MFCD25371360
  • Cas Number : 1429417-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart