PRI424061480 Princeton BioMolecular Research, Inc.

4-Amino-1-(2-fluoro-ethyl)-1H-pyrazole-3-carboxylic acid amide

  • MDL number MFCD25371371
  • Cas Number : 1429419-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart