PRI424061666 Princeton BioMolecular Research, Inc.

2-[(2-methylpyrazole-3-carbonyl)amino]acetic acid

  • MDL number MFCD00462137
  • Molecular Weight : 183.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name {[(1-methyl-1H-pyrazol-5-yl)carbonyl]amino}acetic acid
Inchi Code 1S/C7H9N3O3/c1-10-5(2-3-9-10)7(13)8-4-6(11)12/h2-3H,4H2,1H3,(H,8,13)(H,11,12)
Inchi Key XIGMVYDCHUYWPA-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart