PRI424061694 Princeton BioMolecular Research, Inc.

3-(3-fluorophenyl)-5-methyl-4H-isoxazole-5-carboxylic acid

  • MDL number MFCD19706992
  • Cas Number : 1326815-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart