PRI424061700 Princeton BioMolecular Research, Inc.

4-(2-methoxyphenyl)butanenitrile

  • MDL number MFCD09744580
  • Cas Number : 77853-46-4
  • Molecular Weight : 175.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(2-methoxyphenyl)butanenitrile
Inchi Code 1S/C11H13NO/c1-13-11-8-3-2-6-10(11)7-4-5-9-12/h2-3,6,8H,4-5,7H2,1H3
Inchi Key YQCWMFXWZQZVEC-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart