PRI444785459 Princeton BioMolecular Research, Inc.

[2-(3-Chloro-phenyl)-2-methoxy-ethyl]-methyl-amine

  • MDL number MFCD16681115
  • Cas Number : 1250905-45-3
  • Molecular Weight : 199.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart