PRI444785815 Princeton BioMolecular Research, Inc.

(3-Trifluoromethyl-[1,2,4]triazolo[4,3-b]pyridazin-6-ylsulfanyl)-acetic acid

  • MDL number MFCD15730811
  • Molecular Weight : 278.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart