PRI444786972 Princeton BioMolecular Research, Inc.

(5-Bromo-1-methyl-1H-indazol-3-yloxy)-acetic acid

  • MDL number MFCD20535093
  • Cas Number : 1352532-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature RT
Availability details for
Quantity
  • Availability
  • Add to Cart