PRI444787081 Princeton BioMolecular Research, Inc.

(2S,3R)-2-[[4-(1H-indole-2-carbonyl)piperazine-1-carbonyl]amino]-3-methyl-pentanoic acid

  • MDL number MFCD20535494
  • Molecular Weight : 386.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart