PRI444787887 Princeton BioMolecular Research, Inc.

5-(2,2,2-Trifluoro-ethoxymethyl)-furan-2-carbaldehyde

  • MDL number MFCD22385518
  • Cas Number : 1394022-66-2
  • Molecular Weight : 208.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart