PRI444787899 Princeton BioMolecular Research, Inc.

2,2,5,5-Tetramethyl-tetrahydro-furan-3-ylamine

  • MDL number MFCD19205865
  • Cas Number : 857821-82-0
  • Molecular Weight : 143.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart