PRI444788163 Princeton BioMolecular Research, Inc.

3-Oxa-bicyclo[4.1.0]hept-7-ylamine

  • MDL number MFCD26381465
  • Cas Number : 1559558-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart