PRI444788217 Princeton BioMolecular Research, Inc.

2-Amino-1-phenyl-ethanone

  • MDL number MFCD00064391
  • Cas Number : 613-89-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart