PRI444788333 Princeton BioMolecular Research, Inc.

6-Chloro-8-(2,2-dimethyl-2,3-dihydro-benzofuran-7-yloxymethyl)-4H-benzo[1,3]dioxine

  • MDL number MFCD08741734
  • Molecular Weight : 346.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart