PRI444788411 Princeton BioMolecular Research, Inc.

2-(5-Hydroxy-4-oxo-4H-pyran-2-ylmethylsulfanyl)-benzooxazole-5-carboxylic acid methyl ester

  • MDL number MFCD26097651
  • Molecular Weight : 333.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart