PRI444788983 Princeton BioMolecular Research, Inc.

(4-Cyanomethyl-2-nitro-phenoxy)-acetic acid methyl ester

  • MDL number MFCD28248833
  • Molecular Weight : 250.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart