PRI444789335 Princeton BioMolecular Research, Inc.

2-(3,4-Difluoro-phenyl)-2-methyl-cyclopropylamine

  • MDL number MFCD16744484
  • Molecular Weight : 183.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart