PRI444789488 Princeton BioMolecular Research, Inc.

6-Morpholin-4-ylmethyl-benzo[1,3]dioxol-5-ol

  • MDL number MFCD06886777
  • Molecular Weight : 237.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart