PRI444789489 Princeton BioMolecular Research, Inc.

6-Amino-3-ethyl-1,3-dihydro-indol-2-one

  • MDL number MFCD20705029
  • Cas Number : 23694-70-4
  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart