PRI444789492 Princeton BioMolecular Research, Inc.

4-Aminomethyl-2-nitro-phenylamine

  • MDL number MFCD03410961
  • Cas Number : 79556-69-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart