PRI444789495 Princeton BioMolecular Research, Inc.

6-Aminomethyl-3,4-dihydro-1H-quinolin-2-one

  • MDL number MFCD16987667
  • Cas Number : 933726-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart