PRI444789499 Princeton BioMolecular Research, Inc.

1-Cyclopropyl-5-oxo-pyrrolidine-3-carboxylic acid methyl ester

  • MDL number MFCD23713912
  • Cas Number : 1479122-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart