PRI444789500 Princeton BioMolecular Research, Inc.

6-Aminomethyl-4H-benzo[1,4]oxazin-3-one

  • MDL number MFCD14551328
  • Cas Number : 916303-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart