PRI444789508 Princeton BioMolecular Research, Inc.

(2-chlorophenyl)-cyclopentyl-methanamine

  • MDL number MFCD12824615
  • Cas Number : 1183583-72-3
  • Molecular Weight : 209.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart