PRI444789513 Princeton BioMolecular Research, Inc.

1-[(2-hydroxybenzoyl)amino]-3-methyl-thiourea

  • MDL number MFCD01661561
  • Cas Number : 63224-32-8
  • Molecular Weight : 225.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart