PRI444789514 Princeton BioMolecular Research, Inc.

4-Chloro-2-phenylamino-benzoic acid

  • MDL number MFCD00157007
  • Cas Number : 19218-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart