PRI444789519 Princeton BioMolecular Research, Inc.

1,5-Dimethyl-1H-[1,2,3]triazole-4-carboxylic acid methyl ester

  • MDL number MFCD18830905
  • Cas Number : 175789-79-4
  • Molecular Weight : 155.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart