PRI444789521 Princeton BioMolecular Research, Inc.

(4-Hydroxy-phenylamino)-acetic acid methyl ester

  • MDL number MFCD12159477
  • Cas Number : 247921-34-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart