PRI444789524 Princeton BioMolecular Research, Inc.

3-(4-Ethoxycarbonylamino-phenyl)-acrylic acid

  • MDL number MFCD12107620
  • Molecular Weight : 235.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart