PRI444789528 Princeton BioMolecular Research, Inc.

5-Chloro-thieno[3,2-b]pyridine-3-carbaldehyde

  • MDL number MFCD02571812
  • Cas Number : 152420-81-0
  • Molecular Weight : 197.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart