PRI444789531 Princeton BioMolecular Research, Inc.

2-(3-Bromo-4-methoxy-phenyl)-succinic acid

  • MDL number MFCD01175175
  • Cas Number : 327098-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart