PRI444789532 Princeton BioMolecular Research, Inc.

2-Cyano-3-thiophen-2-yl-thioacrylamide

  • MDL number MFCD01118663
  • Cas Number : 68029-46-9
  • Molecular Weight : 194.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart