PRI444789537 Princeton BioMolecular Research, Inc.

4-Aminomethyl-2-nitro-phenol

  • MDL number MFCD06213143
  • Cas Number : 21078-47-7
  • Molecular Weight : 168.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart