PRI444789544 Princeton BioMolecular Research, Inc.

4-Acetylamino-3-bromo-benzoic acid

  • MDL number MFCD06208852
  • Cas Number : 74103-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart