PRI444789548 Princeton BioMolecular Research, Inc.

[3-(acetoxymethyl)-1,4-dioxido-quinoxaline-1,4-diium-2-yl]methyl acetate

  • MDL number MFCD01106956
  • Cas Number : 10103-89-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart