PRI444789549 Princeton BioMolecular Research, Inc.

5-[4-(Pyrrolidine-1-sulfonyl)-phenyl]-[1,3,4]oxadiazole-2-thiol

  • MDL number MFCD03046986
  • Cas Number : 497926-09-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart